4-Brom-2-fluor-5-iodtoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-3-fluor-6-iod-4-methylbenzol

M-Clarity™ Qualitätsstufe = MQ100

639028-1G
SIAL639028-1GEA 79.2 EUR
SIAL639028-1G
4-Brom-2-fluor-5-iodtoluol, Sigma-Aldrich®
4-Brom-2-fluor-5-iodtoluol
Formel: C₇H₅BrFI
Molecular Weight: 314,92 g/mol
MDL: MFCD06200719
CAS-Nummer: 870704-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR