5-Brom-4-fluor-2-methylanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

639036-5G
SIAL639036-5GEA 85.8 EUR
SIAL639036-5G
5-Brom-4-fluor-2-methylanilin, Sigma-Aldrich®
5-Brom-4-fluor-2-methylanilin
Formel: C₇H₇BrFN
Molecular Weight: 204.04 g/mol
MDL: MFCD05865218
CAS-Nummer: 627871-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR