2-Fluor-5-pyridinboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

639184-1G 639184-5G
SIAL639184-1GEA 22.8 EUR
SIAL639184-1G SIAL639184-5G
2-Fluor-5-pyridinboronsäure, Sigma-Aldrich®
2-Fluor-5-pyridinboronsäure
Formel: C₅H₅BFNO₂
MDL: MFCD03411559
CAS-Nummer: 351019-18-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR