trans-3-Hydroxy-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3-Hydroxyphenyl)-2-nitroethene

M-Clarity™ Qualitätsstufe = MQ100

639672-5G
SIAL639672-5GEA 603 EUR
SIAL639672-5G
trans-3-Hydroxy-β-nitrostyrol, Sigma-Aldrich®
trans-3-Hydroxy-β-nitrostyrol
Formel: C₈H₇NO₃
CAS-Nummer: 3156-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR