trans-2-Hydroxy-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

639680-5G
SIAL639680-5GEA 238 EUR
SIAL639680-5G
trans-2-Hydroxy-β-nitrostyrol, Sigma-Aldrich®
trans-2-Hydroxy-β-nitrostyrol
Formel: C₈H₇NO₃
CAS-Nummer: 3156-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR