trans-2,5-Dimethoxy-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,4-Dimethoxy-2-(2-nitrovinyl)benzene

M-Clarity™ Qualitätsstufe = MQ100

639699-1G
SIAL639699-1GEA 70.8 EUR
SIAL639699-1G
trans-2,5-Dimethoxy-β-nitrostyrol, Sigma-Aldrich®
trans-2,5-Dimethoxy-β-nitrostyrol
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209,200 g/mol
MDL: MFCD00065072
CAS-Nummer: 40276-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR