trans-2-Methoxy-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

639710-1G
SIAL639710-1GEA 33.8 EUR
SIAL639710-1G
trans-2-Methoxy-β-nitrostyrol, Sigma-Aldrich®
trans-2-Methoxy-β-nitrostyrol
Formel: C₉H₉NO₃
MDL: MFCD00070647
CAS-Nummer: 3316-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR