trans-4-Hydroxy-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-4-(2-Nitrovinyl)phenol

M-Clarity™ Qualitätsstufe = MQ100

639729-1G
SIAL639729-1GEA 37.5 EUR
SIAL639729-1G
trans-4-Hydroxy-β-nitrostyrol, Sigma-Aldrich®
trans-4-Hydroxy-β-nitrostyrol
Formel: C₈H₇NO₃
Molecular Weight: 165,1461 g/mol
MDL: MFCD00060551
CAS-Nummer: 22568-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR