4-Amino-1-Boc-piperidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

640042-1G 640042-5G
SIAL640042-1GEA 40.3 EUR
SIAL640042-1G SIAL640042-5G
4-Amino-1-Boc-piperidin, Sigma-Aldrich®
4-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD01076201
CAS-Nummer: 87120-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR