Bis(4-methoxyphenyl)disulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Bis(4-methoxyphenyl)disulphane

M-Clarity™ Qualitätsstufe = MQ100

641367-1G
SIAL641367-1GEA 59.1 EUR
SIAL641367-1G
Bis(4-methoxyphenyl)disulfid, Sigma-Aldrich®
Bis(4-methoxyphenyl)disulfid
Formel: C₁₄H₁₄O₂S₂
Molecular Weight: 278,4 g/mol
MDL: MFCD00041358
CAS-Nummer: 5335-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR