Samarium(III)oxid ∼5% (w/w) ≥99,9% (Metall-Basis), Nanopartikel-Dispersion <100 nm (BET), Sigma-Aldrich®
Lieferant: Merck
641855-100ML
SIAL641855-100MLEA
704
EUR
SIAL641855-100ML
Samarium(III)oxid ∼5% (w/w) ≥99,9% (Metall-Basis), Nanopartikel-Dispersion <100 nm (BET), Sigma-Aldrich®
Samarium(III)oxid
Formel:
O₃Sm₂ Molecular Weight: 348,72 g/mol Schmelzpunkt: 2325 °C Dichte: 7,68 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011237 CAS-Nummer: 12060-58-1 EINECS: 235-043-6 Merck Index: 14,08347 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...