Holmium(III)oxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Holmiumoxid, Diholmiumtrioxid
641863-25ML
SIAL641863-25MLEA
300
EUR
SIAL641863-25ML
Holmium(III)oxid, Sigma-Aldrich®
Holmium(III)oxid
Formel:
Ho₂O₃ Molecular Weight: 377,86 g/mol Siedepunkt: 2600 °C (1013 hPa) Schmelzpunkt: 2400 °C Dichte: 3,79 g/cm³ (20 °C) |
MDL:
MFCD00011053 CAS-Nummer: 12055-62-8 EINECS: 235-015-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...