trans-2,6-Dichlor-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dichloro-β-nitrostyrene

M-Clarity™ Qualitätsstufe = MQ100

642142-1G
SIAL642142-1GEA 32.3 EUR
SIAL642142-1G
trans-2,6-Dichlor-β-nitrostyrol, Sigma-Aldrich®
trans-2,6-Dichlor-β-nitrostyrol
Formel: C₈H₅Cl₂NO₂
Molecular Weight: 218,04 g/mol
MDL: MFCD00024821
CAS-Nummer: 22482-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR