trans-3,4-Dichlor-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4-Dichloro-β-nitrostyrene, 1,2-Dichloro-4-[(E)-2-nitroethenyl]benzene

M-Clarity™ Qualitätsstufe = MQ100

642150-5G
SIAL642150-5GEA 118 EUR
SIAL642150-5G
trans-3,4-Dichlor-β-nitrostyrol, Sigma-Aldrich®
trans-3,4-Dichlor-β-nitrostyrol
Formel: C₈H₅Cl₂NO₂
MDL: MFCD00052834
CAS-Nummer: 18984-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR