trans-4-Chlor-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Chloro-4-(2-nitrovinyl)benzene

M-Clarity™ Qualitätsstufe = MQ100

642177-5G
SIAL642177-5GEA 114 EUR
SIAL642177-5G
trans-4-Chlor-β-nitrostyrol, Sigma-Aldrich®
trans-4-Chlor-β-nitrostyrol
Formel: C₈H₆ClNO₂
MDL: MFCD00024825
CAS-Nummer: 706-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR