trans-2-Chlor-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chloro-β-nitrostyrene, 1-(2-Chlorophenyl)-2-nitroethylene

M-Clarity™ Qualitätsstufe = MQ100

642185-5G
SIAL642185-5GEA 48.8 EUR
SIAL642185-5G
trans-2-Chlor-β-nitrostyrol, Sigma-Aldrich®
trans-2-Chlor-β-nitrostyrol
Formel: C₈H₆ClNO₂
Molecular Weight: 183,60 g/mol
MDL: MFCD00024820
CAS-Nummer: 3156-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR