trans-3-Chlor-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642193-5G
SIAL642193-5GEA 148 EUR
SIAL642193-5G
trans-3-Chlor-β-nitrostyrol, Sigma-Aldrich®
trans-3-Chlor-β-nitrostyrol
Formel: C₈H₆ClNO₂
Molecular Weight: 183,59 g/mol
MDL: MFCD00175518
CAS-Nummer: 37888-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR