trans-3-Brom-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642207-5G
SIAL642207-5GEA 174 EUR
SIAL642207-5G
trans-3-Brom-β-nitrostyrol, Sigma-Aldrich®
trans-3-Brom-β-nitrostyrol
Formel: C₈H₆BrNO₂
CAS-Nummer: 115665-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR