trans-2-Brom-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-1-Bromo-2-(2-nitrovinyl)benzene

M-Clarity™ Qualitätsstufe = MQ100

642215-5G
SIAL642215-5GEA 160 EUR
SIAL642215-5G
trans-2-Brom-β-nitrostyrol, Sigma-Aldrich®
trans-2-Brom-β-nitrostyrol
Formel: C₈H₆BrNO₂
Molecular Weight: 228,0427 g/mol
MDL: MFCD01317567
CAS-Nummer: 155988-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR