3-Brom-p-toluonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642398-25G
SIAL642398-25GEA 72 EUR
SIAL642398-25G
3-Brom-p-toluonitril, Sigma-Aldrich®
3-Brom-p-toluonitril
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
MDL: MFCD06797818
CAS-Nummer: 42872-74-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR