3-Iod-4-methylbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642401-25G
SIAL642401-25GEA 222 EUR
SIAL642401-25G
3-Iod-4-methylbenzonitril, Sigma-Aldrich®
3-Iod-4-methylbenzonitril
Formel: C₈H₆IN
Molecular Weight: 243,444 g/mol
MDL: MFCD06797817
CAS-Nummer: 42872-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR