Methansulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methylsulphonamide

64275-10G 64275-50G
SIAL64275-10GEA 78.5 EUR
SIAL64275-10G SIAL64275-50G
Methansulfonamid, Sigma-Aldrich®
Methansulfonamid
Formel: CH₅NO₂S
MDL: MFCD00007940
CAS-Nummer: 3144-09-0
EINECS: 221-553-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR