2-Amino-3,5-dibrombenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642800-5G
SIAL642800-5GEA 63.7 EUR
SIAL642800-5G
2-Amino-3,5-dibrombenzonitril, Sigma-Aldrich®
2-Amino-3,5-dibrombenzonitril
Formel: C₇H₄Br₂N₂
Molecular Weight: 275,93 g/mol
MDL: MFCD00179571
CAS-Nummer: 68385-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR