2-Amino-3-chlor-5-nitrobenzonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Amino-3-chlor-5-nitrobenzoesäurenitril, 2-chlor-6-cyan-4-nitroanilin
M-Clarity™ Qualitätsstufe = MQ100
642835-25G
SIAL642835-25GEA
142
EUR
SIAL642835-25G
2-Amino-3-chlor-5-nitrobenzonitril, Sigma-Aldrich®
2-Amino-3-chlor-5-nitrobenzonitril
Formel:
C₇H₄ClN₃O₂ Molecular Weight: 197,58 g/mol Schmelzpunkt: 185…187 °C |
MDL:
MFCD00126957 CAS-Nummer: 20352-84-5 EINECS: 243-760-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...