Aluminiumoxid 20% (w/w) in Wasser, Nanopartikel 30 - 60 nm (TEM), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642991-100ML
SIAL642991-100MLEA 320 EUR
SIAL642991-100ML
Aluminiumoxid 20% (w/w) in Wasser, Nanopartikel 30 - 60 nm (TEM), Sigma-Aldrich®
Aluminium oxide

No dispersant is added.

Formel: Al₂O₃
Dichte: 1,06 g/ml (25 °C)
MDL: MFCD00003424
CAS-Nummer: 1344-28-1

Bestellen

Spezifikation Testergebnisse

ICP Major Analysis (Confirms Aluminum Component) Confirmed
Loading, Disperison 19.0 - 21.0% wt.
Particle Size (TEM) 30 - 60 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR