4-Fluor-2-iodtoluol, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 1-Fluor-3-iod-4-methylbenzol, 4-Fluor-2-iod-1-methylbenzol

M-Clarity™ Qualitätsstufe = MQ100

643068-5G
SIAL643068-5GEA 26 EUR
SIAL643068-5G
4-Fluor-2-iodtoluol, Sigma-Aldrich®
4-Fluor-2-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236,03 g/mol
MDL: MFCD00013709
CAS-Nummer: 13194-67-7
EINECS: 236-153-7
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR