3-Chlor-2,4,5-trifluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4,5-Trifluoro-3-chlorobenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

643300-5G
SIAL643300-5GEA 37 EUR
SIAL643300-5G
3-Chlor-2,4,5-trifluorbenzoesäure, Sigma-Aldrich®
3-Chloro-2,4,5-trifluorobenzoic acid
Formel: C₇H₂ClF₃O₂
Molecular Weight: 210,54 g/mol
MDL: MFCD00153101
CAS-Nummer: 101513-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR