2-Acetylamino-3-brom-5-methylpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

643483-1G
SIAL643483-1GEA 63.2 EUR
SIAL643483-1G
2-Acetylamino-3-brom-5-methylpyridin, Sigma-Aldrich®
2-Acetylamino-3-brom-5-methylpyridin
Formel: C₈H₉BrN₂O
Molecular Weight: 229,074 g/mol
MDL: MFCD06798091
CAS-Nummer: 142404-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR