2-Acetylamino-5-brom-4-methylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(5-Bromo-4-methylpyridin-2-yl)acetamide, 2-Acetamido-5-bromo-4-picoline, 2-Acetamido-5-bromo-4-methylpyridine

M-Clarity™ Qualitätsstufe = MQ100

643491-1G
SIAL643491-1GEA 86.4 EUR
SIAL643491-1G
2-Acetylamino-5-brom-4-methylpyridin, Sigma-Aldrich®
2-Acetylamino-5-brom-4-methylpyridin
Formel: C₈H₉BrN₂O
Molecular Weight: 229,0700 g/mol
MDL: MFCD06798092
CAS-Nummer: 142404-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR