2-Acetylamino-5-brom-3-methylpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

643505-1G
SIAL643505-1GEA 50.2 EUR
SIAL643505-1G
2-Acetylamino-5-brom-3-methylpyridin, Sigma-Aldrich®
2-Acetylamino-5-brom-3-methylpyridin
Formel: C₈H₉BrN₂O
Molecular Weight: 229,07 g/mol
MDL: MFCD01928227
CAS-Nummer: 142404-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR