2,6-Dichlor-3-cyan-4-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dichloro-4-methylnicotinonitrile, 2,6-dichloro-4-methylpyridine-3-carbonitrile, 2,6-Dichloro-3-cyano-4-methylpyridine

M-Clarity™ Qualitätsstufe = MQ100

643521-25G
SIAL643521-25GEA 156 EUR
SIAL643521-25G
2,6-Dichlor-3-cyan-4-picolin, Sigma-Aldrich®
2,6-Dichlor-3-cyan-4-picolin
Formel: C₇H₄Cl₂N₂
Molecular Weight: 187.03 g/mol
MDL: MFCD00052633
CAS-Nummer: 875-35-4
EINECS: 212-873-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR