2-Brom-3-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

643548-1G
SIAL643548-1GEA 32.8 EUR
SIAL643548-1G
2-Brom-3-(trifluormethyl)pyridin, Sigma-Aldrich®
2-Bromo-3-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
Molecular Weight: 226 g/mol
MDL: MFCD00153084
CAS-Nummer: 175205-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR