2-Chlor-5-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

643556-5G
SIAL643556-5GEA 82 EUR
SIAL643556-5G
2-Chlor-5-fluorpyridin, Sigma-Aldrich®
2-Chloro-5-fluoropyridine
Formel: C₅H₃ClFN
MDL: MFCD03095332
CAS-Nummer: 31301-51-6
UN: 2929
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR