3-(4-Hydroxyphenyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4'-Hydroxy-[1,1'-biphenyl]-3-carbonitrile, 4-(3-Cyanophenyl)phenol

M-Clarity™ Qualitätsstufe = MQ100

643998-1G
SIAL643998-1GEA 115 EUR
SIAL643998-1G
3-(4-Hydroxyphenyl)benzonitril, Sigma-Aldrich®
3-(4-Hydroxyphenyl)benzonitril
Formel: C₁₃H₉NO
Molecular Weight: 195,2200 g/mol
MDL: MFCD05854255
CAS-Nummer: 154848-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR