(R)-(+)-1-Boc-3-aminopyrrolidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

644064-5G
SIAL644064-5GEA 187 EUR
SIAL644064-5G
(R)-(+)-1-Boc-3-aminopyrrolidin, Sigma-Aldrich®
(R)-(+)-1-Boc-3-aminopyrrolidin
Formel: C₉H₁₈N₂O₂
MDL: MFCD03419272
CAS-Nummer: 147081-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR