DL-Methioninsulfon, Sigma-Aldrich®

Lieferant: Merck

64410-5G
SIAL64410-5GEA 216 EUR
SIAL64410-5G
DL-Methioninsulfon, Sigma-Aldrich®
DL-Methioninsulfon
Formel: C₅H₁₁NO₄S
Molecular Weight: 181,21 g/mol
MDL: MFCD00025079
CAS-Nummer: 820-10-0
EINECS: 212-466-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR