3-Brom-5-formylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromopyridine-5-carboxaldehyde, 5-Bromopyridine-3-carboxaldehyde, 5-Bromonicotinaldehyde, 5-Bromo-3-pyridinecarboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

644102-5G
SIAL644102-5GEA 179 EUR
SIAL644102-5G
3-Brom-5-formylpyridin, Sigma-Aldrich®
3-Brom-5-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
MDL: MFCD03265758
CAS-Nummer: 113118-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR