3'-Nitroacetanilid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-(3-Nitrophenyl)acetamid
M-Clarity™ Qualitätsstufe = MQ100
644250-25G
SIAL644250-25GEA
277
EUR
SIAL644250-25G
3'-Nitroacetanilid, Sigma-Aldrich®
3'-Nitroacetanilid
Formel:
C₈H₈N₂O₃ Dichte: 1 g/cm³ (20 °C) |
MDL:
MFCD00017015 CAS-Nummer: 122-28-1 EINECS: 204-532-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...