Indol-4-carbonitril, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4-Indolcarbonitril, 4-Cyanindol, 1H-Indol-4-carbonitril, 4-Cyan-1H-indol
M-Clarity™ Qualitätsstufe = MQ100
645532-1G
645532-5G
SIAL645532-1GEA
38.2
EUR
SIAL645532-1G
SIAL645532-5G
Indol-4-carbonitril, Sigma-Aldrich®
Indol-4-carbonitril
Formel:
C₉H₆N₂ Molecular Weight: 142.16 g/mol |
MDL:
MFCD00152045 CAS-Nummer: 16136-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...