Indol-4-carbonitril, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4-Indolcarbonitril, 4-Cyanindol, 1H-Indol-4-carbonitril, 4-Cyan-1H-indol

M-Clarity™ Qualitätsstufe = MQ100

645532-1G 645532-5G
SIAL645532-1GEA 38.2 EUR
SIAL645532-1G SIAL645532-5G
Indol-4-carbonitril, Sigma-Aldrich®
Indol-4-carbonitril
Formel: C₉H₆N₂
Molecular Weight: 142.16 g/mol
MDL: MFCD00152045
CAS-Nummer: 16136-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR