Methyl-2,3-dimethoxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3-Dimethoxybenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ200

645583-10G
SIAL645583-10GEA 44.2 EUR
SIAL645583-10G
Methyl-2,3-dimethoxybenzoat, Sigma-Aldrich®
Methyl-2,3-dimethoxybenzoat
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
MDL: MFCD00027555
CAS-Nummer: 2150-42-7
EINECS: 218-425-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR