2-Brom-m-toluidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

645621-5G
SIAL645621-5GEA 322 EUR
SIAL645621-5G
2-Brom-m-toluidin, Sigma-Aldrich®
2-Brom-m-toluidin
Formel: C₇H₈BrN
Molecular Weight: 186,05 g/mol
MDL: MFCD06411367
CAS-Nummer: 54879-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR