2-Fluor-6-iodtoluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

646342-1G
SIAL646342-1GEA 74.6 EUR
SIAL646342-1G
2-Fluor-6-iodtoluol, Sigma-Aldrich®
2-Fluor-6-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236,03 g/mol
MDL: MFCD00142922
CAS-Nummer: 443-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR