1-Brom-4-pentylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brom-pentylbenzol, (4-Bromphenyl)pentan, p-Brom-pentylbenzol

M-Clarity™ Qualitätsstufe = MQ100

646350-5G
SIAL646350-5GEA 73.6 EUR
SIAL646350-5G
1-Brom-4-pentylbenzol, Sigma-Aldrich®
1-Brom-4-pentylbenzol
Formel: C₁₁H₁₅Br
Molecular Weight: 227,14 g/mol
MDL: MFCD00061113
CAS-Nummer: 51554-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR