2-(Boc-amino)-5-brompyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Boc-2-Amino-5-bromopyridine, 2-Amino-5-bromopyridine, 2-BOC protected

M-Clarity™ Qualitätsstufe = MQ100

646385-5G
SIAL646385-5GEA 58.1 EUR
SIAL646385-5G
2-(Boc-amino)-5-brompyridin, Sigma-Aldrich®
2-(Boc-amino)-5-brompyridin
Formel: C₁₀H₁₃BrN₂O₂
Molecular Weight: 273,13 g/mol
MDL: MFCD06411381
CAS-Nummer: 159451-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR