Methyl-3-chlor-2-benzo[b]thiophencarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 3-chlorobenzo[b]thiophene-2-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

646741-10G
SIAL646741-10GEA 257 EUR
SIAL646741-10G
Methyl-3-chlor-2-benzo[b]thiophencarboxylat, Sigma-Aldrich®
Methyl-3-chlor-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₇ClO₂S
Molecular Weight: 226,68 g/mol
MDL: MFCD00052503
CAS-Nummer: 21211-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR