Methyl-3-chlor-6-fluor-2-benzo[b]thiophencarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

646776-1G
SIAL646776-1GEA 110 EUR
SIAL646776-1G
Methyl-3-chlor-6-fluor-2-benzo[b]thiophencarboxylat, Sigma-Aldrich®
Methyl-3-chlor-6-fluor-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₆ClFO₂S
MDL: MFCD00449849
CAS-Nummer: 21211-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR