Methyl-3-chlor-6-methyl-2-benzo[b]thiophencarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

646784-1G
SIAL646784-1GEA 117 EUR
SIAL646784-1G
Methyl-3-chlor-6-methyl-2-benzo[b]thiophencarboxylat, Sigma-Aldrich®
Methyl-3-chlor-6-methyl-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₇ClO₂S
MDL: MFCD01630702
CAS-Nummer: 59812-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR