4-Amino-3-nitropyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine

M-Clarity™ Qualitätsstufe = MQ100

646962-10G
SIAL646962-10GEA 197 EUR
SIAL646962-10G
4-Amino-3-nitropyridin, Sigma-Aldrich®
4-Amino-3-nitropyridin
Formel: C₅H₅N₃O₂
Molecular Weight: 139.11 g/mol
MDL: MFCD00160304
CAS-Nummer: 1681-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR