Methyl-3-brom-4-methoxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brom-4-methoxybenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

647578-10G
SIAL647578-10GEA 145 EUR
SIAL647578-10G
Methyl-3-brom-4-methoxybenzoat, Sigma-Aldrich®
Methyl-3-brom-4-methoxybenzoat
Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
MDL: MFCD00210463
CAS-Nummer: 35450-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR